Methylene blue trihydrate 98.0-103.0% (dried basis), BAKER ANALYZED® C.I. 52015, J.T.Baker®
Catalog # Q473-03
Supplier: Avantor
CAS Number:
Specifications
- Pack type:Glass bottle, amber, wide mouth
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:—
- Solubility:Passes Test
- Identification:Passes Test
- Loss on Drying:8.0 - 18.0 %
- Residue after Ignition:≤ 1.2 %
- Copper (Cu):≤ 0.02 %
- Zinc (Zn):Passes Test
- Assay (C₁₆H₁₈ClN₃S) (dried basis):98.0 - 103.0 %
- Trace Impurities - Arsenic (As):≤ 8.000 ppm
Specification Test Results
About this item
:
:373.9 g/mol
EINECS:200-515-2
:GHS07
Storage Temperature:Ambient
CAS Number:7220-79-3
Formula:C₁₆H₁₈ClN₃S·3H₂O
MDL Number:MFCD00150008