

Diisopropyl ether ≥99.0% (by GC, corrected for water content) stabilised, BAKER ANALYZED® ACS, J.T.Baker®
Catalog # 9243-22
Supplier: Avantor
CAS Number:
Specifications
- Pack type:Aluminium bottle
- Pk:1 L
- Seq:5037
Specifications
Specification Test Results
- Meets ACS Reagent Chemical Requirements:—
- For Laboratory,Research,or Manufacturing Use:= 99.0 %
- Assay ((CH₃)₂CHOCH(CH₃)₂) (by GC, corrected for water):≥99.0 %
- Color (APHA):≤25
- Residue after Evaporation:≤0.0100 %
- Titrable Acid (meq/g):≤0.0007
- Peroxide (as C₆H₁₄O₂):≤0.05 %
Specification Test Results
About this item
:
:102.18 g/mol
Merck Index:13,05232
Boiling Pt:68,5 °C (1013 hPa)
Melting Pt:–86 °C
Density:0,726 g/cm³ (20 °C)
Flash Pt:–28 °C (closed cup)
EINECS:203-560-6
UN:1159
ADR:3,II
:Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol) 0,01 %
:GHS02
:GHS07
Storage Temperature:Ambient
CAS Number:108-20-3
Formula:(CH₃)₂CHOCH(CH₃)₂
MDL Number:MFCD00008880