


Hydrofluoric acid 48,0 - 51,0% (by acidimetry), BAKER ANALYZED® ACS, J.T.Baker®
9560-18
: Avantor
:
- Pk:2,5 L
- Seq:5096
- For Laboratory,Research,or Manufacturing Use:48.0 - 51.0 %
- Assay (HF) (by acidimetry):48.0 - 51.0 %
- Fluosilicic Acid (H₂SiF₆):≤0.01 %
- Residue after Ignition:≤5 ppm
- Chloride (Cl):≤5 ppm
- Nitrate (NO₃):≤5 ppm
- Phosphate (PO₄):≤1 ppm
- Sulfate and Sulfite (as SO₄):≤5 ppm
- Trace Impurities - Arsenic and Antimony (as As):≤50.0 ppb
- Trace Impurities - Copper (Cu):≤50 ppb
- Heavy Metals (as Pb):≤500.0 ppb
- Trace Impurities - Iron (Fe):≤500.0 ppb
- Trace Impurities - Lead (Pb):≤100 ppb
- Trace Impurities - Nickel (Ni):≤50 ppb
:
:20.01 g/mol
:Ambient
:231-634-8
:1790
:8,II
:GHS06
:GHS05
:GHS08
:7664-39-3
:HF
:MFCD00011346