
Ammonium formate ≥96%, J.T.Baker®
Catalog # M530-08
Supplier: Avantor
CAS Number:
Specifications
- Pack type:Poly bottle, white, blue T/E cap
- Pk:1 kg
- Seq:4024
Specifications
Specification Test Results
- Assay (HCOONH4):min. 96%
- Identification (by IR):Passes Test
Specification Test Results
About this item
:
:63.06 g/mol
Merck Index:13,00525
Melting Pt:119…121 °C
Density:1.28 g/cm³ (25 °C)
EINECS:208-753-9
:GHS07
Storage Temperature:Ambient
CAS Number:540-69-2
Formula:HCO₂NH₄
MDL Number:MFCD00013103
Density:1,28 g/cm³ (25 °C)