Order Entry
India
ContactUsLinkComponent
 
 
 
 
Diethyl ether, anhydrous ≥99.0% (by GC, corrected for water content) stabilised, BAKER ANALYZED® ACS suitable for fat extraction, J.T.Baker®
  9244-06
 :  Avantor
 :  
undefined
 
 
 
Diethyl ether, anhydrous ≥99.0% (by GC, corrected for water content) stabilised, BAKER ANALYZED® ACS suitable for fat extraction, J.T.Baker®
  9244-06
 :  Avantor
 :  

 

  • Pack type:
    Aluminium bottle
  • Pk:
    500 mL
  • Seq:
    2725

 

 

  • Meets ACS Reagent Chemical Requirements:
  • For Laboratory,Research,or Manufacturing Use:
    = 99.0 %
  • Assay ((C₂H₅)₂O) (by GC, corrected for water):
    ≥99.0 %
  • Alcohol (C₂H₅OH):
    Passes Test
  • Carbonyl Compounds (as HCHO) (by polarography):
    ≤0.001 %
  • Color (APHA):
    ≤10
  • Peroxide (as H₂O₂):
    ≤1 ppm
  • Preservative (BHT):
    ≥7 ppm
  • Residue after Evaporation:
    ≤0.001 %
  • Titrable Acid (µeq/g):
    ≤0.2
  • Water (by KF, coulometric):
    ≤0.01 %

 

 

 :
 :74.12 g/mol
 :12,03852
 :35 °C (1013 hPa)
 :–123 °C
 :0,7135 g/cm³ (20 °C)
 :–45 °C
 :Ambient
 :200-467-2
 :1155
 :3,I
 :Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol) ≥7 ppm
 :01-2119535785-29
 :GHS02
 :GHS07
 :GHS08
 :60-29-7
 :(CH₃CH₂)₂O
 :MFCD00011646