Order Entry
India
ContactUsLinkComponent
Cobalt(II) nitrate hexahydrate 98.0-102.0% (by EDTA titration), BAKER ANALYZED® ACS, J.T.Baker®
Oxidizing
Corrosive
Health
Environment
Cobalt(II) nitrate hexahydrate 98.0-102.0% (by EDTA titration), BAKER ANALYZED® ACS, J.T.Baker®
Catalog # 1680-01
Supplier:  Avantor
CAS Number:  
undefined
Oxidizing
Corrosive
Health
Environment
Cobalt(II) nitrate hexahydrate 98.0-102.0% (by EDTA titration), BAKER ANALYZED® ACS, J.T.Baker®
Catalog # 1680-01
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    Poly bottle, white, blue T/E cap
  • Pk:
    500 g
  • Seq:
    3645

Specifications

Specification Test Results

  • Meets ACS Reagent Chemical Requirements:
    98.0 - 102.0%
  • For Laboratory,Research,or Manufacturing Use:
    ≤0.01%
  • Assay (Co(NO₃)₂6H₂O) (by EDTA titrn):
    98.0 - 102.0 %
  • Insoluble Matter:
    ≤ 0.01 %
  • pH of 5% Solution at 25°C:
    3.0 - 6.0
  • Chloride (Cl):
    ≤ 0.002 %
  • Sulfate (SO₄):
    ≤ 0.005 %
  • Calcium (Ca):
    ≤ 0.005 %
  • Copper (Cu):
    ≤ 0.002 %
  • Trace Impurities - Iron (Fe):
    ≤ 0.001 %
  • Lead (Pb):
    ≤ 0.002 %
  • Magnesium (Mg):
    ≤ 0.005 %
  • Nickel (Ni):
    ≤ 0.15 %
  • Potassium (K):
    ≤ 0.01 %
  • Sodium (Na):
    ≤ 0.05 %
  • Zinc (Zn):
    ≤ 0.01 %

Specification Test Results

About this item

 :
 :291.03 g/mol
Merck Index:13,02469
Melting Pt:57 °C
Density:1,88 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:233-402-1
UN:1477
ADR:5.1,II
 :GHS03
 :GHS05
 :GHS08
 :GHS09
CAS Number:10026-22-9
Formula:Co(NO₃)₂·6H₂O
MDL Number:MFCD00149647