Order Entry
India
ContactUsLinkComponent
Mag-Bind® blood RNA 96 kit
Mag-Bind® blood RNA 96 kit
Catalog # M2839-00
Supplier:  OMEGA BIOTEK
CAS Number:  
Mag-Bind® blood RNA 96 kit
Catalog # M2839-00
Supplier:  OMEGA BIOTEK
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Method / Format:
    Magnetic beads
  • Prep size:
    Mini
  • Refine target molecule:
    Total RNA
  • Sample type:
    Blood
  • Target molecule:
    RNA
  • Description:
    Mag-Bind® Blood RNA 96 Kit, 200 µl (1×96)
  • Sample size:
    50 - 200 µl
  • Description:
    Mag-Bind® Blood RNA

Specifications

About this item

The Mag-Bind® Blood RNA 96 Kit is designed for rapid and reliable isolation of total and viral RNA from mammalian whole blood. The Mag-Bind® bead technology provides high-quality RNA, which is suitable for direct use in most downstream applications, such as amplifications and enzymatic reactions.