Order Entry
India
ContactUsLinkComponent
E.Z.N.A.® blood RNA kit
E.Z.N.A.® blood RNA kit
Supplier:  OMEGA BIOTEK
Certificates
E.Z.N.A.® blood RNA kit
Supplier:  OMEGA BIOTEK
Certificates

Specifications

  • Method / Format:
    Column (silica-based)
  • Prep size:
    Mini
  • Refine target molecule:
    Total RNA
  • Sample type:
    Blood
  • Target molecule:
    RNA
  • Sample size:
    10 - 1000 µl
  • Description:
    E.Z.N.A.® Blood RNA Kit

Specifications

Related Information

Frequently Bought Together