Wash bottles, wide neck, colour coded, safety labelled, SP Bel-Art
These wash bottles, made of LDPE, translucent, with PP spray cup feature a 53 mm wide mouth for easy refilling and are pre-printed with the chemical name and formula, the National Fire Protection Association (NFPA) section 704 four-colour diamond, health hazards, Chemical Abstract System (CAS) number and suggested protective clothing and equipment. Leak-proof, these bottles have PP polypropylene closures to aid in quick and easy identification.
- Good chemical
- Wide neck for quick, safe filling
- Colour-coded closures for easy identification
Provides compliance with OSHA requirements outlined in Hazard Communication Standard 29 CFR 1910.1200.
: 500 ml bottles are packaged as 4/bag; 6 bags/case.
Frequently Bought Together