Order Entry
India
ContactUsLinkComponent
 
Water with 0,1% (v/v) trifluoroacetic acid, BAKER ANALYZED® for LC-MS, J.T.Baker®
  9836-03
 :  Avantor
 :  
undefined
Water with 0,1% (v/v) trifluoroacetic acid, BAKER ANALYZED® for LC-MS, J.T.Baker®
  9836-03
 :  Avantor
 :  

 

  • Pack type:
    Glass bottle, amber, narrow mouth
  • Pk:
    4 L
  • Seq:
    5162

 

 

  • For Laboratory,Research,or Manufacturing Use:
  • Appearance:
    Passes Test
  • Trifluoroacetic Acid:
    0.095 - 0.105 %
  • Gradient Elution Test (a.u.) - 254 nm:
    ≤0.01
  • Residue after Evaporation:
    ≤1.0 ppm
  • LC/MS Suitability - Largest Response on ESI- Positive Mode (as Reserpine):
    ≤50 ppb
  • Trace Impurities - Aluminum (Al):
    ≤50.0 ppb
  • Trace Impurities - * Calcium (Ca):
    ≤50.0 ppb
  • Trace Impurities - Iron (Fe):
    ≤50.0 ppb
  • Trace Impurities - Magnesium (Mg):
    ≤50.0 ppb
  • Trace Impurities - Potassium (K):
    ≤50 ppb
  • Trace Impurities - * Sodium (Na):
    ≤50.0 ppb
  • * May change over time due to extraction from glass container.:
  • Filtered through a 0.2 micron filter.:

 

 

 :
 :Ambient
 :200-929-3
 :76-05-1