Order Entry
India
ContactUsLinkComponent
L(+)-Arabinose ≥99%, high purity
L(+)-Arabinose ≥99%, high purity
Supplier:  VWR Chemicals
Certificates
undefined
L(+)-Arabinose ≥99%, high purity
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Arsenic:
    <0.0003%
  • Assay (dry basis):
    ≥99%
  • Chloride:
    ≤0.005%
  • Heavy Metals (as Pb):
    ≤0.0001%
  • Loss on Drying:
    ≤0.5%
  • Melting Point:
    154 - 158 °C
  • Residue on Ignition:
    ≤0.1%
  • Specific Rotation:
    101 - 105°
  • Sulphate:
    ≤0.005%

Specification Test Results

Related Information

Frequently Bought Together