Order Entry
India
ContactUsLinkComponent
Corning® Matrigel® hESC-Qualified Matrix, Corning
Corning® Matrigel® hESC-Qualified Matrix, Corning
Catalog # 734-1440
Supplier:  Corning
   Low Manufacturing Impact
CAS Number:  
undefined
Corning® Matrigel® hESC-Qualified Matrix, Corning
Catalog # 734-1440
Supplier:  Corning
   Low Manufacturing Impact
CAS Number:  

Specifications

  • Pk:
    5 mL
  • Description:
    Corning® Matrigel® matrix, hESC-qualified
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    ISO 14001 - Bedford

Specifications

About this item

Pre-screned for compatibility with mTeSR™1 medium by StemCell Technologies providing the reproducibility and consistency essential for hESC and iPSC feeder-free stem cell culture.

  • Mouse colonies routinely screened for pathogens via Mouse Antibody Production (MAP) testing
  • Extensive PCR testing is performed to screen for a number of pathogens
  • Tested and found negative for bacteria, fungi, and mycoplasma
  • Protein concentrations determined by Lowry method
  • Endotoxin units measured by Limulus Amoebocyte Lysate assay
  • Matrigel Matrix gel stability tested for a period of 14 days at 37°C
  • Biological activity determined for each lot using a neurite outgrowth assay.

Corning Matrigel matrix is a solubilized basement membrane preparation extracted from the Engelbreth-Holm-Swarm (EHS) mouse sarcoma, a tumor rich in extracellular matrix proteins, including Laminin (a major component), Collagen IV, heparin sulfate proteoglycans, entactin/nidogen, and a number of growth factors.

Application
• Cell Growth and Differentiation
• Metabolism/Toxicology Studies
• Invasion Assays
• In Vitro and In Vivo Angiogenesis Assays
• In Vivo Angiogenesis Studies and Augmentation of Tumors in Immunosuppressed Mice

Preparation and Storage
Store at -20°C. Avoid multiple freeze-thaws