Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
Rs.0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
India
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
Standard solution imidazole 1000 mg/l in water
Standard solution imidazole 1000 mg/l in water
:
VWR Chemicals
Standard solution imidazole 1000 mg/l in water
:
VWR Chemicals
CAS No.:
288-32-4
Formula:
C₃H₄N₂
Description:
Standard solution imidazol
sds_VWRC87345
VWR® for Calibrations
Frequently Bought Together
VWR®, Cone and Plate viscosity standards
: VWR Chemicals
Water Reag. Ph. Eur. 1095507 particle-free
: VWR Chemicals
VWR® AVS TITRINORM Buffer Solutions, 20 °C
: VWR Chemicals
Lead acetate solution Reag. Ph. Eur. 1048103
: VWR Chemicals
VWR®, N,N-Dimethylformamide, secondary reference standard for GC, PESTINORM®
: VWR Chemicals
VWR®, Sucrose (Brix) standards, stabilised solutions for refractometry
: VWR Chemicals
VWR®, Conductivity standard solutions
: VWR Chemicals
pH buffer, single-use, SINGLET™
: HACH