APS/TEMED, tablets for biotechnology
Catalog # N310-100TAB
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:100 Tab.
- Seq:0940
Specifications
Specification Test Results
- Dissolution Time:<= 4 minutes
- Polymerization Time:<= 30 minutes
- Solubility (1 tablet/1mL Water):PASS
- Tablet Weight:112 - 137 mg
Specification Test Results
About this item
:
UN:1479
ADR:5.1,III
:GHS07
:GHS08
:GHS05
:GHS03
Storage Temperature:Refrigerator
CAS Number:7727-54-0/110-18-9
:110-18-9