Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
Rs.0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
India
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Hydroxylamine
Hydroxylamine solution, alkaline (I+II) Reag. Ph. Eur. 1044302
Hydroxylamine solution, alkaline (I+II) Reag. Ph. Eur. 1044302
87848.290
:
VWR Chemicals
:
Hydroxylamine solution, alkaline (I+II) Reag. Ph. Eur. 1044302
87848.290
:
VWR Chemicals
:
Pack type:
Plastic bottle
Pk:
1 L
Seq:
5037
:
:
33.03 g/mol
:
7803-49-8
:
H₃NO
:
MFCD00044522