Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
Rs.0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
India
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Cupri-tartaric solution
Cupri-tartaric solution R4 (I+II) Reag. Ph. Eur. 1023304
Cupri-tartaric solution R4 (I+II) Reag. Ph. Eur. 1023304
Cupri-tartaric solution R4 (I+II) Reag. Ph. Eur. 1023304
Catalog # 87815.290
Supplier:
VWR Chemicals
CAS Number:
SDS
Certificates
Expand image
Cupri-tartaric solution R4 (I+II) Reag. Ph. Eur. 1023304
Catalog # 87815.290
Supplier:
VWR Chemicals
CAS Number:
SDS
Certificates
Specifications
Pack type:
Plastic bottle
Pk:
1 L
Seq:
5037
Specifications
About this item
Product Details
Documentation
:
Product Details
Documentation