Bacterial RNA kit, E.Z.N.A.®
Supplier: OMEGA BIOTEK
Certificates
About this item
The E.Z.N.A.® Bacterial RNA kit is designed for isolation of high-quality total RNA from variety of bacterial strains. Up to 1×10⁹ log-phase bacterial cells can be processed. This kit uses an improved lysis procedure to ensure the complete lysis of bacterial cells. Purified RNA is suitable for downstream applications such as RT-PCR and hybridisation techniques.
- Optimised buffers guarantee pure RNA every time
- No organic extractions
- Purified RNA suitable for most applications
Specifications
- Method / Format:Column (silica-based)
- Prep size:Mini
- Refine target molecule:Bacterial RNA
- Sample type:Bacteria
- Target molecule:RNA
- Sample size:Up to 1×109 bacteria
- Description:E.Z.N.A.® Bacterial RNA Kit
Specifications
Frequently Bought Together