Order Entry
India
ContactUsLinkComponent
Phenol : chloroform : iso-amyl alcohol (25:24:1) (v/v/v) stabilised
Toxic
Corrosive
Health
Phenol : chloroform : iso-amyl alcohol (25:24:1) (v/v/v) stabilised
Catalog # A2279.0500
Supplier:  PanReac AppliChem
CAS Number:  
Toxic
Corrosive
Health
Phenol : chloroform : iso-amyl alcohol (25:24:1) (v/v/v) stabilised
Catalog # A2279.0500
Supplier:  PanReac AppliChem
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Seq:
    2725

Specifications

About this item

 :
ADR:6.1,II
 :Stabilised with 8-hydroxyquinoline
UN:2810
 :GHS06
 :GHS05
 :GHS08
Storage Temperature:Ambient
CAS Number:136112-00-0
MDL Number:MFCD00133763