Order Entry
India
ContactUsLinkComponent
E.Z.N.A.® stool DNA kit
E.Z.N.A.® stool DNA kit
Supplier:  OMEGA BIOTEK
Certificates
undefined
E.Z.N.A.® stool DNA kit
Supplier:  OMEGA BIOTEK
Certificates

Specifications

  • Method / Format:
    Column (silica-based)
  • Prep size:
    Mini
  • Refine target molecule:
    Total DNA
  • Sample type:
    Stool
  • Target molecule:
    DNA
  • Sample size:
    200 mg
  • Description:
    E.Z.N.A.® Stool DNA Kit

Specifications

Related Information

Frequently Bought Together