VWR® Mixing Measuring Cylinders, Tall Form, Class A, Borosilicate Glass
Supplier: VWR Collection
Certificates
About this item
Borosilicate glass 3,3, class A.
- Round base with interchangeable PE stopper
Certifications: DIN 12685, ISO 4788, with LOT certificate.
Frequently Bought Together