VWR® Measuring Cylinders, Tall Form, Class A, Borosilicate Glass
Supplier: VWR Collection
Certificates
About this item
Borosilicate glass 3,3, graduated, class A.
- Hexagonal glass base, with spout
- Serially numbered
Certifications: Individual work certificate, DIN 12680.
Frequently Bought Together