VWR® Filter Holders for Microanalysis
Supplier: VWR Collection
Certificates
About this item
Application: It is used to filter under vacuum/small volumes for particulate or biological contamination analysis. Use durapore (PVDF) membrane or unlaminated PTFE for solvent applications with this apparatus.
- Filter is available in fritted glass or stainless steel
- Thick prefilters or laminated filters will not seal in this holder
Specifications
- Catalog No:
- 511-0205
- 511-0199
- Filtration area:
- 2,5
- 2,5