VWR® Reagent Bottles, Narrow Neck, with Screw Cap
215-3173
- Ext.Ø:56 mm
- Capacity:100 ml
- Height:100 mm
- Seq:01
- Pk:10
Borosilicate glass, grade 3.3.
With autoclavable screw cap and pouring ring manufactured from ASTM E438 Type 1 Class A.
: In compliance with DIN ISO 4796 - 1 standards.