Order Entry
India
Orders LinkContactUsLinkComponent
VWR® Erlenmeyer Flasks, Conical, with Interchangable Joint
250 ml, 185 mm, 24/29 NS
  214-0312
VWR® Erlenmeyer Flasks, Conical, with Interchangable Joint
  214-0312
VWR® Erlenmeyer Flasks, Conical, with Interchangable Joint
250 ml, 185 mm, 24/29 NS
1 * 10 items
Borosilicate glass, grade 3.3.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
The good value alternative for a wide range of laboratory applications. Volume graduations and labelling area.
 : DIN 12387
  • Socket size:
    24/29 NS
  • Capacity:
    250 ml
  • Height:
    185 mm
  • Seq:
    01