Order Entry
India
ContactUsLinkComponent
VWR® Beakers, Low Form, Borosilicate Glass
VWR® Beakers, Low Form, Borosilicate Glass
Catalog # 213-0363
Supplier:  VWR Collection
Supplier #:
undefined
VWR® Beakers, Low Form, Borosilicate Glass
Catalog # 213-0363
Supplier:  VWR Collection
Supplier #:
Log in for availability
Select an Option
83 mm
Select an Option
500 ml
Select an Option
115 mm
Select an Option
08
Select an Option
8
About this item
Borosilicate glass, grade 3.3.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
With spout and graduation.
Certifications: DIN 12331, ISO 3819
Frequently Bought Together