VWR® Flasks, Flat Bottom, Single Neck, with Interchangeable Joint
Supplier: VWR Collection
Certificates
About this item
Borosilicate glass, grade 3.3.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
The inexpensive alternative for a wide range of laboratory applications. With matt labelling area. Imprinted catalogue number for convenient reordering.
Certifications: DIN 12348
Frequently Bought Together