VWR® Protein Molecular Weight Marker, Wide Range
Catalog # K494-500UL
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pk:500 µl
- Description:Protein MW markers, wide range
Specifications
About this item
Protein molecular weight marker, wide range offers eight bands ranging from 14,4 to 212,0 kDa. There is sufficient material for 100 to 150 assays.
- 8 bands ranging from 14,4 to 212,0 kDa
- Ideal for high or low molecular weight applications