VWR® Prism Protein Molecular Weight Markers
Catalog # 1B1349-500UL
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pk:500 µl
- Description:Prism protein marker
Specifications
About this item
The Prism protein marker contains six purified proteins stained orange, blue, and violet for easy molecular weight estimation of samples. The bands ranging from 19 to 130 kDa can simplify monitoring protein migration during electrophoresis, and subsequently assess protein transfer efficiency to a blotting membrane. Prism protein marker is ready to load and may be applied to lab-prepared or commercial pre-cast gels. Sufficient material for 50 to 100 lanes.
- Six bands ranging from 19 to 130 kDa
- Three vibrant colours for easy band identification