Tetrabutylammonium dihydrogen phosphate for HPLC, J.T.Baker®
Catalog # V375-03
Supplier: Avantor
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:= 99.0 %
- Assay (C₁₆H₃₈NO₄P):≥99.0 %
- Ultraviolet Absorbance (0.1% Solution in Water) - 196 nm:≤1.00
- Ultraviolet Absorbance (0.1% Solution in Water) - 200 nm:≤0.20
- Ultraviolet Absorbance (0.1% Solution in Water) - 210 nm:≤0.15
- Ultraviolet Absorbance (0.1% Solution in Water) - 220-400 nm:≤0.01
- Appearance (white to off-white powder):Passes Test
- pH of 5% Solution at 25°C:3.8 - 4.5
Specification Test Results
About this item
:
:339.46 g/mol
EINECS:226-947-1
CAS Number:5574-97-0
Formula:C₁₆H₃₈NO₄P
MDL Number:MFCD00064526