Lanthanum(III) chloride heptahydrate 64,5 - 70,0% (by anhydrous basis), BAKER ANALYZED® ACS, J.T.Baker®
Supplier: Avantor
Certificates
About this item
CAS Number:10025-84-0
:
Formula:LaCl₃·7H₂O
MDL Number:MFCD00149756
Merck Index:13,05379
Boiling Pt:860 °C (1013 hPa)
Melting Pt:857 °C
Density:3,8773 g/cm³ (–0.15 °C)
:371.37 g/mol
EINECS:233-237-5
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Meets ACS Reagent Chemical Requirements:64.5 - 70.0%
- For Laboratory,Research,or Manufacturing Use:≤0.01%
- ACS - Assay (LaCl₃):64.5 - 70.0 %
- ACS - Insoluble Matter:≤0.01 %
- ACS - Calcium (Ca):≤0.001 %
- Trace Impurities - Magnesium (Mg):≤10 ppm
Specification Test Results
Frequently Bought Together