Chromium(III) chloride hexahydrate 99.0-105.0% (by iodometry), BAKER ANALYZED®, J.T.Baker®
Supplier: Avantor
Certificates
About this item
CAS Number:10060-12-5
:
Formula:CrCl₃·6H₂O
MDL Number:MFCD00149660
:266.45 g/mol
Merck Index:13,02242
Melting Pt:85 °C
Density:1,76 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:233-038-3
UN:3260
ADR:8,III
:GHS07
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:99.0 - 105.0 %
- Assay (CrCl₃6H₂O) (by iodometry):99.0 - 105.0 %
- Insoluble Matter:≤ 0.01 %
- pH of 5% Solution at 25°C:2.0 - 3.0
- Sulfate (SO₄):≤ 0.01 %
- Iron (Fe):≤ 0.01 %
Specification Test Results
Frequently Bought Together