


Nitrogen monoxide cylinder, emptied/unwashed
Catalog # 600723S.EMPTY
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pk:15 L
- Seq:5246
Specifications
About this item
:
Merck Index:12,06674
EINECS:233-271-0
UN:1660
ADR:2,
:GHS03
:GHS06
:GHS05
CAS Number:10102-43-9
Formula:NO