Order Entry
India
Orders LinkContactUsLinkComponent
millipore_microscopy_feature.jpg
Harleco & Midas - Made for Each Other
Microscopy products to support your success.
    Related Information:
    Clinical microscopy | Avantor
    millipore_dyes_stains_100.jpg
    • Highest Quality Stain Solutions - IVD certified, also certified by the Biological Stain Commission
    • Ready to use staining solutions for blood and bone marrow samples with intense and reproducible results
    • Very clean results in less than 1 minute with Hemacolor®
    Learn more
    millipore_midas_100.jpg
    The Midas® III-Plus Automated Stainer
    • The six station unit holds 20 or 30 slides
    • Up to 10 programs - each program can have up to 29 steps
    • Save up to 50% on stain consumption vs. flood or spray techniques
    Learn more