Order Entry
India
ContactUsLinkComponent
mflx-pump-tubing-1920x760.jpg
Masterflex® Peristaltic Pump Tubing
Accept no substitutes — Precision extruded & pump tested formulations engineered to provide the highest quality for tubing life, pressure, and suction lift.
Masterflex brand pump tubing is custom extruded to fit Masterflex pumps and engineered for long life in peristaltic pump applications. Lot-to-lot consistency provides accuracy and repeatability.
undefined
Masterflex Pump Tubing Selection Guide
The right pump tubing is crucial when building your fluid processing pump system. Before you choose, consider all aspects of your application: chemical compatibility, operating temperature, pressure, necessary regulatory approvals, gas permeability of the tubing, and cleaning/sterilization requirements.
Select tubing
puri-flex-siloxane-replacement-300x120.jpg
Is the global siloxane shortage affecting you?
We have you covered without alternative tubing solutions
Learn more
NOTE: PharmaPure and Chem-Durance Bio tubing formulations are co-extruded with different inner and outer materials and require modified testing methods. Contact Masterflex®, part of Avantor for chemical testing instructions. Check our Chemical Resistance Database.
How to verify your tubing compatibility
mflx-tubing-test-step1.jpg
Weigh and Measure the length and diameter of tubing sample
Other featured categories
Top Tubing Formulations
Custom services and tools
Shop by pump tubing series
Need more information or help selecting the perfect product for your application?
Our experts are ready to help. Contact us at 1-800-MASTERFLEX or [email protected]
Contact us