Order Entry
India
Orders LinkContactUsLinkComponent
cmsitem_00571142_CLIA_Hero.jpg
CLIA
BeneSphera™ (A Brand of Avantor) - CLIA Product Range
CLIA | Avantor
Introducing BeneSphera™ Chemiluminescence Assay Product Range – Illuminate diagnostics with our advanced assays, delivering unparalleled sensitivity and accuracy. Explore a new era in clinical testing, where precision meets innovation for superior patient care.
Chemiluminescence assays are important in various scientific and medical fields due to their unique characteristics and applications.
  • High Sensitivity
  • Wide Dynamic Range
  • Low Background Noise
  • Multiplexing Capability
  • Automation and High Throughput
Ad1_cmsitem_00571142_CLIA_.jpg
CL-1000i is a robust and simple-to-use chemiluminescence analyzer with a throughput of up to 120 tests per hour. In a compact benchtop package, it has a large onboard capacity of 25 reagents and 60 samples, and supports non-stop refill of samples, reagents and consumables during testing. By integrating the reliable CLIA technology with latest electrical and software improvement, CL-1000i enables easy operation and brings confident diagnosis faster than ever.
See the product
Ad2_cmsitem_00571142_CLIA_.jpg
As one of the world’s smallest, fully automated chemiluminescence immunoassay analyzers, the CL-900i integrates a large capacity and fast assay speed into a compact model, achieving a perfect balance between size and immunoassay testing performance.
See the product
Ad3_cmsitem_00571142_CLIA_.jpg
Maglumi 800, is a chemiluminescence Immunoassay (CLIA) System with a throughput of 180 tests per hour, capable of performing more than 166 parameters.
See the product
Ad4_cmsitem_00571142_CLIA_.jpg
The MAGLUMI@ X3 fully automatic analyzer is designed for quantitative and qualitative in-vitro diagnosis. Targeted for small and middle-size labs or hospitals, also a good choice as a backup instrument for big labs or hospitals, the compact design of MAGLUMI X3 significantly reduces analyzer size and weight, but still maintains the throughput of 200 tests per hour.
See the product