Order Entry
India
Orders LinkContactUsLinkComponent
Hero_cmsitem_00571141.jpg
5 Part Analyzers and Advance Analyzers
BeneSphera™ (A Brand of Avantor) - 3 Part Hematology Analyzers
5 Part Analyzers and Advance Analyzers | Avantor
Ad1_cmsitem_00571141.jpg
The BC 5150 Auto Hematology Analyzer is a light and compact system, specially tailored to assist diagnostic labs who need full CBC+5 part results, with a relatively low daily sample volume. Responding to increased demands for high quality read-outs and accurate diagnostic decisions, the BC 5150 provides a reliable complete 5-part differential menu, 25 reportable parameters and 12 research parameters, 3 histograms and 3 scattergrams.
See the product
Ad2_cmsitem_00571141.jpg
With the newly designed optics and reagent systems, the SF Cube technology can help doctors to better differentiate the clusters of cells, which is the key to revealing more abnormal cells
See the product