Order Entry
Ireland
Orders LinkContactUsLinkComponent
Hydranal-Reagents=955.jpg
Hydranal™ Reagents for water determination
Hydranal™ Reagents for water determination | Avantor
Water content plays an important role in the performance of many products. Hydranal by Honeywell Research Chemicals is your trusted provider of the complete portfolio of reagents for water determination by Karl Fischer Titration. Hydranal reagents and water standards have always been developed and produced in our plant in Seelze, Germany, meaning you can be sure to enjoy the same composition, quality, service and technical support you always have. The next generation in Karl Fischer titration is here.
titrants-and-reagent-260.jpg
Explore Honeywell's broad product range of high-quality titrants and reagents for volumetric applications. Advantages of Hydranal Reagents:
  • High titration speed
  • Stable end points
  • Accurate results
  • Long shelf life
  • Wide applicability
  • World leading technical support
Find our products
Coulometric-Titration-260.jpg
Wide range of coulometric reagents based on different solvent compositions serve to support the broad variety of samples analyzed. Advantages of Hydranal Coulometric Reagents:
  • Easy to use
  • High accuracy for trace amounts of water
  • Stable conditions of the titration vessel
  • Broad product range
  • Long shelf life (up to five years)
Find our products
Water-standards-260.jpg
Quality management plays an important role in Karl Fischer (KF) titration. Advance your analysis and research with Hydranal Water Standards and CRMs:
  • Broad product range for volumetric and coulometric KF titration, as well as the indirect oven application
  • Manufactured according to current ISO requirements
  • Verified against external standards such as NIST SRM 2890
  • Long shelf life
  • Convenient packaging with detailed instruction for use
  • Documentation of Analysis showing the exact water content and expiry date provided with each package
Find our products
frequently-260.jpg
Tips and tricks to perform and avoid mistakes in this complex method
Download the document