Order Entry
Ireland
ContactUsLinkComponent
MyCapLC kit containing components to assemble three complete 1/16 OD x 150 µm ID x 50 mm length UHPLC columns, with 250 mm inlet an d 100 mm outlet tails 360 µm OD x 50 µm ID 1 * 3 items
Catalog # SGEA2251110
New Product
MyCapLC kit containing components to assemble three complete 1/16 OD x 150 µm ID x 50 mm length UHPLC columns, with 250 mm inlet an d 100 mm outlet tails 360 µm OD x 50 µm ID 1 * 3 items
Catalog # SGEA2251110