Order Entry
Ireland
ContactUsLinkComponent
1/16 GC ferrule 0.4 mm ID (long) three-hole 85% Vespel / 15% Graphite PK10 1 * 10 items
Catalog # SGEA215194
New Product
1/16 GC ferrule 0.4 mm ID (long) three-hole 85% Vespel / 15% Graphite PK10 1 * 10 items
Catalog # SGEA215194