Order Entry
Ireland
ContactUsLinkComponent
ActivArmr 27806 - Heavy-duty supported nitrile gloves, offering superb abrasion, grease and oil resistance - Fully dipped nitrile coating: Excellent barrier against oil and grease - 100% cotton fleece liner: Optimal wearer comfort assured - Abrasion 1 * 1 PAIR
Catalog # ANSE27806110
Supplier:  Ansell
New Product
ActivArmr 27806 - Heavy-duty supported nitrile gloves, offering superb abrasion, grease and oil resistance - Fully dipped nitrile coating: Excellent barrier against oil and grease - 100% cotton fleece liner: Optimal wearer comfort assured - Abrasion 1 * 1 PAIR
Catalog # ANSE27806110
Supplier:  Ansell