Order Entry
Ireland
ContactUsLinkComponent
Autoclavable Tyvek Breather Bag, With Easy Chevron Peel Feature & Heat Seal Closure, With Steam Indicator Dot, Size: 3 x 6 1 * 1.000 items
Catalog # KACC00306TWBBSD
New Product
Autoclavable Tyvek Breather Bag, With Easy Chevron Peel Feature & Heat Seal Closure, With Steam Indicator Dot, Size: 3 x 6 1 * 1.000 items
Catalog # KACC00306TWBBSD