Order Entry
Ireland
ContactUsLinkComponent
Human MICB ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination o f human MICB in serum, plasma, tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA270824-48
Supplier:  ANTIBODIES.COM
New Product
Human MICB ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination o f human MICB in serum, plasma, tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA270824-48
Supplier:  ANTIBODIES.COM