Order Entry
Ireland
ContactUsLinkComponent
Rat AVPR1B ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination o f rat AVPR1B in tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA270949-48
Supplier:  ANTIBODIES.COM
New Product
Rat AVPR1B ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination o f rat AVPR1B in tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA270949-48
Supplier:  ANTIBODIES.COM