Order Entry
Ireland
ContactUsLinkComponent
Llama Interferon gamma Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detect ion antibody, Streptavidin-AP, recombinant llama Interferon gamma ELISA standard, and standard reconstitution buffer. 1 * 1.920 Tests
Catalog # ANTIA270422-2096
Supplier:  ANTIBODIES.COM
New Product
Llama Interferon gamma Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detect ion antibody, Streptavidin-AP, recombinant llama Interferon gamma ELISA standard, and standard reconstitution buffer. 1 * 1.920 Tests
Catalog # ANTIA270422-2096
Supplier:  ANTIBODIES.COM