Order Entry
Ireland
ContactUsLinkComponent
Human FOXO1A ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human FOXO1A in tissue homogenates, cell lysates or other biological fluids. 1 * 96 Tests
  ANTIA2689-96
New Product
Human FOXO1A ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human FOXO1A in tissue homogenates, cell lysates or other biological fluids. 1 * 96 Tests
  ANTIA2689-96