Order Entry
Ireland
ContactUsLinkComponent
Rat IL-17B ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination o f rat IL-17B in serum, plasma or other biological fluids. 1 * 48 Tests
Catalog # ANTIA2766-48
Supplier:  ANTIBODIES.COM
New Product
Rat IL-17B ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination o f rat IL-17B in serum, plasma or other biological fluids. 1 * 48 Tests
Catalog # ANTIA2766-48
Supplier:  ANTIBODIES.COM