Order Entry
Ireland
ContactUsLinkComponent
Human AKT2 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination o f human AKT2 in tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
  ANTIA2776-48
New Product
Human AKT2 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination o f human AKT2 in tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
  ANTIA2776-48