Order Entry
Ireland
ContactUsLinkComponent
Mouse Prosurfactant Protein C ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitat ive determination of mouse Prosurfactant Protein C in serum, plasma, tissue homogenates, cell lysates, lung lavage fl 1 * 96 Tests
Catalog # ANTIA270673-96
Supplier:  ANTIBODIES.COM
New Product
Mouse Prosurfactant Protein C ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitat ive determination of mouse Prosurfactant Protein C in serum, plasma, tissue homogenates, cell lysates, lung lavage fl 1 * 96 Tests
Catalog # ANTIA270673-96
Supplier:  ANTIBODIES.COM