Order Entry
Ireland
ContactUsLinkComponent
Human GTF2A1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human GTF2A1 in serum, plasma, tissue homogenates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA2452-96
Supplier:  ANTIBODIES.COM
New Product
Human GTF2A1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human GTF2A1 in serum, plasma, tissue homogenates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA2452-96
Supplier:  ANTIBODIES.COM