Order Entry
Ireland
ContactUsLinkComponent
Human KLF10 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human KLF10 in tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
  ANTIA2271-48
New Product
Human KLF10 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human KLF10 in tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
  ANTIA2271-48